Fmoc-D-Asp(OtBu)-(Hmb)Gly-OH
Supplier: BACHEM AMERICAS INC
4035340.0001
4035340.0005
B-3675.0001BAEA
1290.81
CAD
B-3675.0001BA
B-3675.0005BA
Fmoc-D-Asp(OtBu)-(Hmb)Gly-OH
Proteins and Peptides
Please see also the corresponding amino acid derivatives Fmoc-(Dmb)Ala-OH (B-4175), Fmoc-(Dmb)Gly-OH (B-4170), Fmoc-(Hmb)Gly-OH (B-3490), and Fmoc-(Dmb)Leu-OH (B-4165).
A building block for the synthesis of peptides containing the D-Asp-Gly motif avoiding base-catalyzed aspartimide formation.