Fmoc-α-Me-L-Leu-OH

Supplier: BACHEM AMERICAS INC

4040839.0001 4040839.0005
B-3665.0001BAEA 1032.67 CAD
B-3665.0001BA B-3665.0005BA
Fmoc-α-Me-L-Leu-OH
Fmoc-alpha-Me-L-Leu-OH

Sequence: Fmoc-α-Me-Leu-OH


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₂₂H₂₅NO₄
Storage Temperature: Freezer
MDL Number: MFCD08458572
CAS Number: 312624-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR