Fmoc-D-Asp(Ompe)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-((3-methylpentan-3-yl)oxy)-4-oxobutanoic acid

4039622.0001 4039622.0005
B-3655.0001BAEA 1004.2 CAD
B-3655.0001BA B-3655.0005BA
Fmoc-D-Asp(Ompe)-OH
Fmoc-D-Asp(Ompe)-OH

The OMpe-side chain protecting group significantly reduces aspartimide formation, an undesired side-chain reaction encountered with the repetitive piperidine treatment needed for Fmoc removal.


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


The OMpe-side chain protecting group significantly reduces aspartimide formation, an undesired side-chain reaction encountered with the repetitive piperidine treatment needed for Fmoc removal.

Formula: C₂₅H₂₉NO₆
MW: 439.51 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08458576
CAS Number: 1926162-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR