Galnon Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4040530.0025 4040530.0100
B-3645.0025BAEA 335.58 CAD
B-3645.0025BA B-3645.0100BA
Galnon Trifluoroacetate
Proteins and Peptides
The galanin receptor agonist galnon binds to the same brain-surface proteins, or receptors, that galanin does and prevents seizures. This drug mimicking a natural substance in the brain may represent a prototype for a new class of epilepsy drugs.