8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-NH-PEG2-CH2COOH, 8-(Fmoc-amino)-3,6-dioxa-n-octanoic acid, Fmoc-8-amino-3,6-dioxaoctanoic acid, [2-[2-[(Fmoc-amino)ethoxy]ethoxy]acetic acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, [2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]acetic acid

4037914.0500 4037914.1000 4037914.5000
B-3635.0500BAEA 335.58 CAD
B-3635.0500BA B-3635.1000BA B-3635.5000BA
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid

Sequence: Fmoc-AEEAc-OH


See also Biotinyl-AEEAc-AEEAc-OH, G-4740.


Synonym(s): Fmoc-Adoa-OH#8-(Fmoc-amino)-3,6-dioxa-octanoic acid#{2-[2-(Fmoc-amino)-ethoxy]-ethoxy}-acetic acid#Fmoc-8-amino-3,6-dioxa-octanoic acid

Formula: C₂₁H₂₃NO₆
MW: 385.42 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01321015
CAS Number: 166108-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR