Fmoc-Glu(Edans)-OH

Supplier: BACHEM AMERICAS INC

4033590.0500 4033590.1000
B-3620.0500BAEA 1086.93 CAD
B-3620.0500BA B-3620.1000BA
Fmoc-Glu(Edans)-OH
Fmoc-Glu(Edans)-OH

Sequence: Fmoc-Glu(EDANS)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Building block for the synthesis of internally quenched fluorogenic peptide substrates (FRET substrates). The fluorescent group EDANS can be efficiently quenched by the nonfluorescent DABCYL chromophore.

Formula: C₃₂H₃₁N₃O₈S
Storage Temperature: Freezer
MDL Number: MFCD01076138
CAS Number: 193475-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR