(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tritylthio)butanoic acid

Supplier: BACHEM AMERICAS INC

4039317.0001 4039317.0005
B-3610.0001BAEA 1419.9 CAD
B-3610.0001BA B-3610.0005BA
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tritylthio)butanoic acid
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tritylthio)butanoic acid

Sequence: Fmoc-D-Homocys(Trt)-OH


The cysteine homolog is formed by S-demethylation of methionine. Contrary to Cys, Hcy can be incorporated and derivatized without risk of racemization and β-elimination. Base-catalyzed desulfuration of peptides containing a Cys/Hcy mixed disulfide bridge yields cystathionine (carbacystine) peptides.


Synonym(s): (R)-2-(Fmoc-amino)-4-tritylsulfanyl-butyric acid

Formula: C₃₈H₃₃NO₄S
Storage Temperature: Freezer
MDL Number: MFCD05663440
CAS Number: 1007840-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR