Fmoc-Arg(Z)₂-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Arg(Z)2-OH

4003522.0001 4003522.0005
B-3600.0001BAEA 516.34 CAD
B-3600.0001BA B-3600.0005BA
Fmoc-Arg(Z)₂-OH
Fmoc-Arg(Z)₂-OH

Sequence: Fmoc-Arg(Z)₂-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₃₇H₃₆N₄O₈
Storage Temperature: Freezer
MDL Number: MFCD05663439
CAS Number: 207857-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR