Boc-AGL -OH

Supplier: BACHEM AMERICAS INC

4039208.0500 4039208.1000
B-3585.0500BAEA 3100.72 CAD
B-3585.0500BA B-3585.1000BA
Boc-AGL -OH
Boc-AGL -OH

Sequence: Fmoc-α-amino-D-Gly(Boc)-OH


Bachem offers derivatives of α-aminoglycine, cyclohexylglycine, cyclopentylglycine, and the PNA building block N-β-aminoethylglycine. For t-butylglycine derivatives see t-Leucine.


Synonym(s): Fmoc-D-Agl(Boc)-OH#Boc-α-amino-Gly(Fmoc)-OH#(R)-2-(Boc-amino)-2-(Fmoc-amino)-acetic acid#Boc-Agl(Fmoc)-OH

Formula: C₂₂H₂₄N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD05663438
CAS Number: 306773-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR