4-(Fmoc-amino)-1-methyl-1H-pyrrole-2-carboxylic acid

Supplier: BACHEM AMERICAS INC

4039215.0250 4039215.1000
B-3580.0250BAEA 851.92 CAD
B-3580.0250BA B-3580.1000BA
4-(Fmoc-amino)-1-methyl-1H-pyrrole-2-carboxylic acid
4-(Fmoc-amino)-1-methyl-1H-pyrrole-2-carboxylic acid

Sequence: 4-(Fmoc-amino)-1-methyl-1H-pyrrole-2-carboxylic acid


Building block for the synthesis of pyrrole-imidazole polyamides. These polyamides can be designed to bind to specific DNA sequences and have been shown to be able to modulate gene expression. Please see also the imidazole derivative B-3410.

Formula: C₂₁H₁₈N₂O₄
MW: 362.385 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02094546
CAS Number: 195387-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR