Fmoc-Asp(Ompe)-OH

Supplier: BACHEM AMERICAS INC

4033988.0001 4033988.0005
B-3560.0001BAEA 727.94 CAD
B-3560.0001BA B-3560.0005BA
Fmoc-Asp(Ompe)-OH
Fmoc-Asp(Ompe)-OH

Sequence: Fmoc-Asp(OMpe)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


The OMpe (O-3-methylpent-3-yl) side chain protecting group significantly reduces aspartimide formation, an undesired side-chain reaction encountered with the repetitive piperidine treatment needed for Fmoc removal.

Formula: C₂₅H₂₉NO₆
Storage Temperature: Freezer
MDL Number: MFCD10001333
CAS Number: 180675-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR