Fmoc-Lys(Ac)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-acetyl-L-lysine, Nalpha-Fmoc-Nepsilon-acetyl-L-lysine, Nepsilon-Acetyl-Nα-Fmoc-L-lysine, Fmoc-Lys(Ac)-OH
4034189.0001
4034189.0005
4034189.0025
B-3520.0001BAEA
197.47
CAD
B-3520.0001BA
B-3520.0005BA
B-3520.0025BA
Fmoc-Lys(Ac)-OH
Fmoc-Lys(Ac)-OH
Sequence: Fmoc-Lys(Ac)-OH
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Formula:
C₂₃H₂₆N₂O₅ MW: 410.47 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00077409 CAS Number: 159766-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...