Fmoc-Lys(Ddiv)-OH

Supplier: BACHEM AMERICAS INC

4034540.0001 4034540.0005
B-3515.0001BAEA 585.22 CAD
B-3515.0001BA B-3515.0005BA
Fmoc-Lys(Ddiv)-OH
Fmoc-Lys(Ddiv)-OH

Sequence: Fmoc-Lys(ivDde)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


The ivDde protective group possesses higher orthogonality regarding Boc and Fmoc in comparison to Dde.

Formula: C₃₄H₄₂N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD01631658
CAS Number: 204777-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR