Fmoc-N(Hmb)-Gly-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-(Hmb)Gly-OH

4034334.0001 4034334.0005
B-3490.0001BAEA 903.62 CAD
B-3490.0001BA B-3490.0005BA
Fmoc-N(Hmb)-Gly-OH
Fmoc-N(Hmb)-Gly-OH

Sequence: Fmoc-(Hmb)Gly-OH


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.


Synonym(s): Fmoc-N-(2-hydroxy-4-methoxy-Bzl)-Gly-OH

Formula: C₂₅H₂₃NO₆
MW: 433.46 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03093500
CAS Number: 148515-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR