Fmoc-L-leucinol

Supplier: BACHEM AMERICAS INC

4033337.0001 4033337.0005 4033337.0025
B-3485.0001BAEA 180.91 CAD
B-3485.0001BA B-3485.0005BA B-3485.0025BA
Fmoc-L-leucinol
Fmoc-L-leucinol

Sequence: Fmoc-L-leucinol

Formula: C₂₁H₂₅NO₃
Storage Temperature: Freezer
MDL Number: MFCD00235932
CAS Number: 139551-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR