Fmoc-O-(benzylphospho)-L-threonine

Supplier: BACHEM AMERICAS INC

4029346.0001 4029346.0005
B-3460.0001BAEA 651.39 CAD
B-3460.0001BA B-3460.0005BA
Fmoc-O-(benzylphospho)-L-threonine
Fmoc-O-(benzylphospho)-L-threonine

Sequence: Fmoc-Thr(PO(OBzl)OH)-OH


Bachem also offers phosphothreonine derivatives.
Phosphothreonine is a potential N-terminal site for chemical ligation.


Building block for the synthesis of phosphothreonine-containing peptides. Harris et al. observed a considerable extent of S-benzylation of adjacent Met residues during microwave-assisted TFA cleavage.

Formula: C₂₆H₂₆NO₈P
Storage Temperature: Freezer
MDL Number: MFCD00797870
CAS Number: 175291-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR