Fmoc-Nα-methyl-L-tryptophan

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-N-Me-Trp-OH

4029285.0250 4029285.1000
B-3430.0250BAEA 828.77 CAD
B-3430.0250BA B-3430.1000BA
Fmoc-Nα-methyl-L-tryptophan
Fmoc-Nalpha-methyl-L-tryptophan

Sequence: Fmoc-N-Me-Trp-OH


MeTrp, αMeTrp.

Formula: C₂₇H₂₄N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD02684512
CAS Number: 112913-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR