Fmoc-N-Me-Ser(tBu)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Fmoc-O-tert-butyl-N-methyl-L-serine
4029103.0001
4029103.0005
B-3400.0001BAEA
850.04
CAD
B-3400.0001BA
B-3400.0005BA
Fmoc-N-Me-Ser(tBu)-OH
Fmoc-N-Me-Ser(tBu)-OH
Sequence: Fmoc-N-Me-Ser(tBu)-OH
Bachem also offers derivatives of phosphoserine and sulfoserine.
Formula:
C₂₃H₂₇NO₅ MW: 397.471 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02094430 CAS Number: 197632-77-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...