Fmoc-His(pi-Me)-OH

Supplier: BACHEM AMERICAS INC

4028985.0250 4028985.1000 4028985.5000
B-3365.0250BAEA 335.58 CAD
B-3365.0250BA B-3365.1000BA B-3365.5000BA
Fmoc-His(pi-Me)-OH
Fmoc-His(pi-Me)-OH

Sequence: Fmoc-His(3-Me)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Fmoc-His(π-Me)-OH

Formula: C₂₂H₂₁N₃O₄
Storage Temperature: Freezer
MDL Number: MFCD02259494
CAS Number: 252049-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR