(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(2,3-dimethylguanidino)pentanoic acid

Supplier: BACHEM AMERICAS INC

4028831.0001 4028831.0005
B-3345.0001BAEA 1626.45 CAD
B-3345.0001BA B-3345.0005BA
(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(2,3-dimethylguanidino)pentanoic acid
(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(2,3-dimethylguanidino)pentanoic acid

Sequence: Fmoc-Arg(Me)₂-OH (symmetrical)


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also E-2770, H-Arg(Me)-OH · HCl, in the subfamily 'Arginine Substrates and Inhibitors'.


Synonym(s): Fmoc-SDMA hydrochloride salt

Formula: C₂₃H₂₈N₄O₄
Storage Temperature: Freezer
MDL Number: MFCD02684483
CAS Number: 823780-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR