Fmoc-L-4-methylphe

Supplier: BACHEM AMERICAS INC

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methyl-L-phenylalanine, N-Fmoc-4-methyl-L-phenylalanine, Fmoc-Phe(4-Me)-OH, Fmoc-p-Me-Phe-OH

4028886.0001 4028886.0005
B-3335.0001BAEA 387.21 CAD
B-3335.0001BA B-3335.0005BA
Fmoc-L-4-methylphe
Fmoc-L-4-methylphe

Sequence: Fmoc-p-Me-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Formula: C₂₅H₂₃NO₄
MW: 401.462 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270197
CAS Number: 199006-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR