Fmoc-N-(2-boc-aminoethyl)-Gly-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-N-(N-β-Boc-aminoethyl)-Gly-OH, Fmoc-N-(2-Boc-aminoethyl)-Gly-OH

4028556.0001 4028556.0005
B-3285.0001BAEA 903.62 CAD
B-3285.0001BA B-3285.0005BA
Fmoc-N-(2-boc-aminoethyl)-Gly-OH
Fmoc-N-(2-boc-aminoethyl)-Gly-OH

Sequence: Fmoc-N-(N-β-Boc-aminoethyl)-Gly-OH


Bachem offers derivatives of α-aminoglycine, cyclohexylglycine, cyclopentylglycine, and the PNA building block N-β-aminoethylglycine. For t-butylglycine derivatives see t-Leucine.


Fmoc-Aeg(Boc)-OH, key building block in combination with an allyl-type anchor in the solid-phase synthesis of PNAs.

Formula: C₂₄H₂₈N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD02259490
CAS Number: 141743-15-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR