Fmoc-Chg-OH

Supplier: BACHEM AMERICAS INC

4028499.0001 4028499.0005 4028499.0025
B-3270.0001BAEA 232.35 CAD
B-3270.0001BA B-3270.0005BA B-3270.0025BA
Fmoc-Chg-OH
Fmoc-Chg-OH

Sequence: Fmoc-cyclohexyl-Gly-OH


Bachem offers derivatives of α-aminoglycine, cyclohexylglycine, cyclopentylglycine, and the PNA building block N-β-aminoethylglycine. For t-butylglycine derivatives see t-Leucine.
Chg derivatives.


Synonym(s): Fmoc-Chg-OH

Formula: C₂₃H₂₅NO₄
Storage Temperature: Freezer
MDL Number: MFCD00190878
CAS Number: 161321-36-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR