Fmoc-Glu(OAll)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-L-glutamic acid 5-allyl ester, N-Fmoc-L-glutamic acid 5-Allyl ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic acid 5-Allyl ester, Fmoc-Glu(allyl ester)-OH, 5-Allyl N-Fmoc-L-glutamate, Fmoc-Glu(OAll)-OH, 5-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate

4028191.0005 4028191.0025
B-3255.0005BAEA 820.34 CAD
B-3255.0005BA B-3255.0025BA
Fmoc-Glu(OAll)-OH
Fmoc-Glu(OAll)-OH

Sequence: Fmoc-Glu(allyl ester)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Synonym(s): Fmoc-Glu(OAll)-OH

Formula: C₂₃H₂₃NO₆
MW: 409.438 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00190879
CAS Number: 133464-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR