Fmoc-N-Me-Nle-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Fmoc-N-methyl-L-norleucine
4028697.0001
4028697.0005
B-3230.0001BAEA
309.81
CAD
B-3230.0001BA
B-3230.0005BA
Fmoc-N-Me-Nle-OH
Fmoc-N-Me-Nle-OH
Sequence: Fmoc-N-Me-Nle-OH
Synonym(s): Fmoc-N-Me-L-2-aminohexanoic acid
Formula:
C₂₂H₂₅NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00235878 CAS Number: 112883-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...