Fmoc-N-methyl-L-norvaline
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-N-Me-Nva-OH, N-Fmoc-N-methyl-L-norvaline, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-norvaline, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)pentanoic acid
4028701.0001
4028701.0005
B-3225.0001BAEA
309.81
CAD
B-3225.0001BA
B-3225.0005BA
Fmoc-N-methyl-L-norvaline
Fmoc-N-methyl-L-norvaline
Sequence: Fmoc-N-Me-Nva-OH
Synonym(s): Fmoc-N-Me-L-2-aminovaleric acid
Formula:
C₂₁H₂₃NO₄ MW: 353.418 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01861326 CAS Number: 252049-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...