(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(((4-methoxyphenyl)diphenylmethyl)amino)hexanoic acid

Supplier: BACHEM AMERICAS INC

4028286.0001 4028286.0005
B-3215.0001BAEA 361.31 CAD
B-3215.0001BA B-3215.0005BA
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(((4-methoxyphenyl)diphenylmethyl)amino)hexanoic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(((4-methoxyphenyl)diphenylmethyl)amino)hexanoic acid

Sequence: Fmoc-Lys(4-methoxytrityl)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Synonym(s): Fmoc-Lys(Mmt)-OH

Formula: C₄₁H₄₀N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD00270542
CAS Number: 159857-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR