Fmoc-D-4-fluorophe

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-Phe(4-F)-OH, Fmoc-p-fluoro-D-Phe-OH, N-Fmoc-4-fluoro-D-phenylalanine

4028285.0001 4028285.0005
B-3210.0001BAEA 370.51 CAD
B-3210.0001BA B-3210.0005BA
Fmoc-D-4-fluorophe
Fmoc-D-4-fluorophe

Sequence: Fmoc-p-fluoro-D-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Formula: C₂₄H₂₀FNO₄
MW: 405.425 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00235900
CAS Number: 177966-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR