Fmoc-D-Glu(oall)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-Glu(allyl ester)-OH, Fmoc-D-Glu(OAll)-OH

4028168.0001 4028168.0005
B-3205.0001BAEA 601.51 CAD
B-3205.0001BA B-3205.0005BA
Fmoc-D-Glu(oall)-OH
Fmoc-D-Glu(oall)-OH

Sequence: Fmoc-D-Glu(allyl ester)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Synonym(s): Fmoc-D-Glu(OAll)-OH

Formula: C₂₃H₂₃NO₆
MW: 409.438 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01074690
CAS Number: 204251-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR