Fmoc-Orn(Dde)-OH

Supplier: BACHEM AMERICAS INC

4027859.0001 4027859.0005
B-3185.0001BAEA 569.87 CAD
B-3185.0001BA B-3185.0005BA
Fmoc-Orn(Dde)-OH
Fmoc-Orn(Dde)-OH

Sequence: Fmoc-Orn(Dde)-OH


Bachem also offers Fmoc-Orn(Boc)-Wang resin (D-1800).


The Nδ-protecting group Dde is orthogonal to both Fmoc and Boc which allows on-resin modification of ornithine side chains during Fmoc/tBu-based SPPS. The applications of Dde side-chain protection include synthesis of branched or di-epitopic peptides, preparation of MAP core molecules and lipo-MAPS, the synthesis of cyclic peptides, TASP molecules, and templates for combinatorial chemistry and synthetic proteins. Dde can be cleaved by 2% hydrazine in DMF.

Formula: C₃₀H₃₄N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD01862895
CAS Number: 269062-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR