Fmoc-Lys(Retro-Abz-Boc)-OH

Supplier: BACHEM AMERICAS INC

4027855.0250 4027855.1000
B-3180.0250BAEA 542.13 CAD
B-3180.0250BA B-3180.1000BA
Fmoc-Lys(Retro-Abz-Boc)-OH
Fmoc-Lys(Retro-Abz-Boc)-OH

Sequence: Fmoc-Lys(retro-Abz-Boc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Bachem also offers resins preloaded with Nε-labeled Fmoc-lysine: Fmoc-Lys(biotinyl)-Wang resin (D-2705) and Fmoc-Lys(Dnp)-SASRIN resin (D-2450).


Synonym(s): Boc-Abz-(Fmoc-Lys-OH)#Fmoc-ablysin(Boc)-OH

Formula: C₃₃H₃₇N₃O₇
MW: 587.673 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01318747
CAS Number: 159322-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR