Fmoc-L-4,4'-biphenylalanine

Supplier: BACHEM AMERICAS INC

4027843.0001 4027843.0005
B-3155.0001BAEA 828.77 CAD
B-3155.0001BA B-3155.0005BA
Fmoc-L-4,4'-biphenylalanine
Fmoc-L-4,4'-biphenylalanine

Sequence: Fmoc-4-phenyl-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
4-Bip derivatives.


Synonym(s): Fmoc-β-(4-biphenylyl)-Ala-OH#Fmoc-Bip-OH

Formula: C₃₀H₂₅NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00191198
CAS Number: 199110-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR