Fmoc-Leu-Gly-OH

Supplier: BACHEM AMERICAS INC

4015473.0001 4015473.0005
B-3150.0001BAEA 464.57 CAD
B-3150.0001BA B-3150.0005BA
Fmoc-Leu-Gly-OH
Proteins and Peptides

Fmoc-Leu-Gly can self-assemble forming ultrathin hydrogel membranes.

Order Now

SPECIFICATIONS

Protein/peptide name Fmoc-Leu-Gly-OH
Environmentally Preferable
Species
Source
CAS No. 82007-05-4
MDL No. MFCD01318744
Sequence Fmoc-Leu-Gly-OH
Molecular weight 410.47
Molecular formula C₂₃H₂₆N₂O₅
Storage conditions RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR