Fmoc-D-Asp(OBzl)-OH

Supplier: BACHEM AMERICAS INC

4027692.0001 4027692.0005 4027692.0025
B-3135.0001BAEA 309.81 CAD
B-3135.0001BA B-3135.0005BA B-3135.0025BA
Fmoc-D-Asp(OBzl)-OH
Fmoc-D-Asp(OBzl)-OH

Sequence: Fmoc-D-Asp(OBzl)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₂₆H₂₃NO₆
Storage Temperature: Ambient
MDL Number: MFCD00273445
CAS Number: 150009-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR