Fmoc-S-Boc-3-aminopropyl-L-cysteine
Supplier: BACHEM AMERICAS INC
4028163.0001
4028163.0005
B-3120.0001BAEA
903.62
CAD
B-3120.0001BA
B-3120.0005BA
Fmoc-S-Boc-3-aminopropyl-L-cysteine
Fmoc-S-Boc-3-aminopropyl-L-cysteine
Sequence: Fmoc-Cys(3-(Boc-amino)-propyl)-OH
This protected trifunctional amino acid derivative, representing an analog of homolysine, can be used for introducing a lysine or arginine mimetic. It has been incorporated in ⁹⁹Tc-chelating peptides.
Formula:
C₂₆H₃₂N₂O₆S Storage Temperature: Ambient |
MDL Number:
MFCD01318742 CAS Number: 173963-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...