Fmoc-D-allo-Thr-OH

Supplier: BACHEM AMERICAS INC

4027554.0001 4027554.0005
B-3090.0001BAEA 903.62 CAD
B-3090.0001BA B-3090.0005BA
Fmoc-D-allo-Thr-OH
Fmoc-D-allo-Thr-OH

Sequence: Fmoc-D-allo-Thr-OH


Bachem also offers phosphothreonine derivatives.
aThr derivatives.


Building block for the SPPS of kahalalide F. The hydroxyl group of D-aThr is involved in the ester bond of this cyclic depsipeptide.

Formula: C₁₉H₁₉NO₅
Storage Temperature: Ambient
MDL Number: MFCD01318738
CAS Number: 130674-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR