Fmoc-D-allo-threoninol

Supplier: BACHEM AMERICAS INC

4027547.0001 4027547.0005
B-3075.0001BAEA 774.5 CAD
B-3075.0001BA B-3075.0005BA
Fmoc-D-allo-threoninol
Fmoc-D-allo-threoninol

Sequence: Fmoc-D-allo-threoninol

Formula: C₁₉H₂₁NO₄
Storage Temperature: Freezer
MDL Number: MFCD01318736
CAS Number: 143143-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR