Fmoc-p-carboxy-phe(OtBu)-OH

Supplier: BACHEM AMERICAS INC

4027514.0001 4027514.0005
B-3070.0001BAEA 828.52 CAD
B-3070.0001BA B-3070.0005BA
Fmoc-p-carboxy-phe(OtBu)-OH
Fmoc-p-carboxy-phe(OtBu)-OH

Sequence: Fmoc-p-carboxy-Phe(OtBu)-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Formula: C₂₉H₂₉NO₆
Storage Temperature: Freezer
MDL Number: MFCD01317016
CAS Number: 183070-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR