Fmoc-D-CIS-HYP-OH

Supplier: BACHEM AMERICAS INC

4027501.0001 4027501.0005
B-3065.0001BAEA 828.77 CAD
B-3065.0001BA B-3065.0005BA
Fmoc-D-CIS-HYP-OH
Fmoc-D-CIS-HYP-OH

Sequence: Fmoc-D-cis-Hyp-OH


Hyp denotes L-trans-hydroxyproline.

Formula: C₂₀H₁₉NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD01318735
CAS Number: 214852-45-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR