Fmoc-D-Lys(Fmoc)-OH

Supplier: BACHEM AMERICAS INC

4027404.0001 4027404.0005 4027404.0025
B-3050.0001BAEA 175.66 CAD
B-3050.0001BA B-3050.0005BA B-3050.0025BA
Fmoc-D-Lys(Fmoc)-OH
Fmoc-D-Lys(Fmoc)-OH

Sequence: Fmoc-D-Lys(Fmoc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₃₆H₃₄N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00270743
CAS Number: 75932-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR