(Fmoc-Cys-OtBu)₂

Supplier: BACHEM AMERICAS INC

4027211.0001 4027211.0005
B-3030.0001BAEA 258.14 CAD
B-3030.0001BA B-3030.0005BA
(Fmoc-Cys-OtBu)₂
(Fmoc-Cys-OtBu)₂

Sequence: (Fmoc-Cys-OtBu)₂


As the disulfide bond can be cleaved under mild conditions (e.g. with trialkylphosphines) cystine derivatives are highly economic educts for synthesizing Cys derivatives. The cystine derivatives of fluorophores (e.g. J-1175) or the pNA derivatives may be chosen as educts for synthesizing Cys substrates.


Precursor of Fmoc-Cys-OtBu which has been used in the synthesis of orthogonally protected lanthionines. Barany et al. employed Fmoc-Cys-OtBu in developing a side-chain anchoring strategy for the SPPS of peptide acids with C-terminal cysteine.

Formula: C₄₄H₄₈N₂O₈S₂
Storage Temperature: Refrigerator
MDL Number: MFCD08458535
CAS Number: 139592-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR