Fmoc-3-(1-naphthyl)-D-alanine
Supplier: BACHEM AMERICAS INC
4027577.0001
4027577.0005
B-3020.0001BAEA
464.74
CAD
B-3020.0001BA
B-3020.0005BA
Fmoc-3-(1-naphthyl)-D-alanine
Fmoc-3-(1-naphthyl)-D-alanine
Sequence: Fmoc-D-1-Nal-OH
1-Nal and 2-Nal have been used as Trp replacements.
Formula:
C₂₈H₂₃NO₄ Storage Temperature: Freezer |
MDL Number:
MFCD00191200 CAS Number: 138774-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...