Fmoc-D-Cys(4-methoxytrityl)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-S-((4-methoxyphenyl)diphenylmethyl)-D-cysteine

4027076.0250 4027076.1000
B-2990.0250BAEA 516.34 CAD
B-2990.0250BA B-2990.1000BA
Fmoc-D-Cys(4-methoxytrityl)-OH
Fmoc-D-Cys(4-methoxytrityl)-OH

Sequence: Fmoc-D-Cys(4-methoxytrityl)-OH
Synonym(s): Fmoc-D-Cys(Mmt)-OH


Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).


4-Methoxytrityl (Mmt) is a very acid sensitive cysteine protecting group that can selectively be removed by 0.5 - 1.0% TFA in the presence of protecting groups of the t-butyl type and S-Trt. It can be used in the solid-phase synthesis of cysteine-containing peptides, and further to selectively protect a thiol function in order to introduce a labeling group.

Formula: C₃₈H₃₃NO₅S
MW: 615.75 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02094091
CAS Number: 1198791-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR