N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-phenylacetic acid, N-Fmoc-L-α-phenylglycine, Fmoc-Phg-OH, N-Fmoc-L-2-phenylglycine
4026939.0001
4026939.0005
4026939.0025
B-2980.0001BAEA
180.91
CAD
B-2980.0001BA
B-2980.0005BA
B-2980.0025BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine
Sequence: Fmoc-Phg-OH
Formula:
C₂₃H₁₉NO₄ MW: 373.41 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00155632 CAS Number: 102410-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...