Fmoc-D-Hyp(tBu)-OH

Supplier: BACHEM AMERICAS INC

4027773.0001 4027773.0005
B-2975.0001BAEA 1807.14 CAD
B-2975.0001BA B-2975.0005BA
Fmoc-D-Hyp(tBu)-OH
Fmoc-D-Hyp(tBu)-OH

Sequence: Fmoc-D-Hyp(tBu)-OH


Hyp denotes L-trans-hydroxyproline.

Formula: C₂₄H₂₇NO₅
MW: 409.48 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798649
CAS Number: 268729-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR