Fmoc-D-Dap(Boc)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid
4027406.0001
4027406.0005
B-2965.0001BAEA
570.65
CAD
B-2965.0001BA
B-2965.0005BA
Fmoc-D-Dap(Boc)-OH
Fmoc-D-Dap(Boc)-OH
Sequence: Fmoc-D-Dap(Boc)-OH
Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.
Bachem also offers Fmoc-Dap(Dnp)-SASRIN resin (D-2590) for the SPPS of FRET substrates.
Formula:
C₂₃H₂₆N₂O₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00798648 CAS Number: 198544-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...