Fmoc-D-Dab(Boc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid

4027196.0001 4027196.0005
B-2960.0001BAEA 774.5 CAD
B-2960.0001BA B-2960.0005BA
Fmoc-D-Dab(Boc)-OH
Fmoc-D-Dab(Boc)-OH

Sequence: Fmoc-D-Dab(Boc)-OH

Formula: C₂₄H₂₈N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00798647
CAS Number: 114360-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR