Fmoc-D-Asp(OPp)-OH
Supplier: BACHEM AMERICAS INC
4027583.0001
4027583.0005
B-2955.0001BAEA
800.3
CAD
B-2955.0001BA
B-2955.0005BA
Fmoc-D-Asp(OPp)-OH
Fmoc-D-Asp(OPp)-OH
Sequence: Fmoc-D-Asp(2-phenylisopropyl ester)-OH
Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.
Synonym(s): Fmoc-D-Asp(OPp)-OH
Formula:
C₂₈H₂₇NO₆ Storage Temperature: Freezer |
MDL Number:
MFCD00798646 CAS Number: 214852-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...