Fmoc-D-Asp-ODmb

Supplier: BACHEM AMERICAS INC

4027652.0001 4027652.0005
B-2945.0001BAEA 748.45 CAD
B-2945.0001BA B-2945.0005BA
Fmoc-D-Asp-ODmb
Fmoc-D-Asp-ODmb

Sequence: Fmoc-D-Asp-ODmb


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₂₈H₂₇NO₈
Storage Temperature: Freezer
MDL Number: MFCD00798644
CAS Number: 200335-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR