N-Fmoc-D-norleucine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-Nle-OH

4027625.0005 4027625.0025
B-2925.0005BAEA 284.04 CAD
B-2925.0005BA B-2925.0025BA
N-Fmoc-D-norleucine
N-Fmoc-D-norleucine

Sequence: Fmoc-D-Nle-OH
Synonym(s): Fmoc-D-2-aminohexanoic acid


Bachem also offers Fmoc-Nle-Wang resin (D-1795) and Fmoc-Nle-SASRIN resin (D-1730).

Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00155639
CAS Number: 112883-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR